2,3,4-Trimethoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14278-30
A14278-09
A14278-18
CAAAAA14278-30EA
808.96
CAD
CAAAAA14278-30
CAAAA14278-09
CAAAA14278-18
2,3,4-Trimethoxybenzoic acid 98+%
2,3,4-Trimethoxybenzoic acid
Purity: 98+%
CAS No.: 573-11-5
Molecular Formula: C10H12O5
Formula Weight: 212.20
Odor: Odorless
Melting Point: 99-102°
Beilstein Reference: 2696073
MDL No.: MFCD00002433
Formula:
C₁₀H₁₂O₅ MW: 212.2 g/mol Boiling Pt: 110…115 °C (0,04 mmHg) Melting Pt: 99…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002433 CAS Number: 573-11-5 EINECS: 209-350-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...