2-Adamantanone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Adamantan-2-one

A14275-06 A14275-14 A14275-22
CAAAA14275-06EA 79.16 CAD
CAAAA14275-06 CAAAA14275-14 CAAAA14275-22
2-Adamantanone 98%
2-Adamantanone

Purity: 98% CAS No.: 700-58-3 Molecular Formula: C10H14O Formula Weight: 150.22 Melting Point: ca 257° subl. Beilstein Reference: 1210235

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 228
Melting Pt: 43 (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3
EINECS: 211-847-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR