3,4,5-Trimethoxybenzoic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14274-22
A14274-36
CAAAA14274-22EA
114.87
CAD
CAAAA14274-22
CAAAA14274-36
3,4,5-Trimethoxybenzoic acid 99+%
3,4,5-Trimethoxybenzoic acid
Purity: 99+%
CAS No.: 118-41-2
Molecular Formula: C10H12O5
Formula Weight: 212.20
Melting Point: 168-171°
Boiling Point: 225-226°/10mm
MDL No.: MFCD00002501
Formula:
C₁₀H₁₂O₅ Boiling Pt: 225…226 °C (10 mmHg) Melting Pt: 168…171 °C Density: 1,31 g/cm³ (24.95 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002501 CAS Number: 118-41-2 EINECS: 204-248-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...