1-Phenyl-1-propyne 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14272-06
A14272-09
CAAAAA14272-06EA
109.18
CAD
CAAAAA14272-06
CAAAA14272-09
CAAAA14272-18
1-Phenyl-1-propyne 98%
1-Phenyl-1-propyne
Purity: 98%
CAS No.: 673-32-5
Molecular Formula: C9H8
Formula Weight: 116.16
Storage Temperature: 2°C to 8°C
Flash Point: 62°(143°F)
Refractive Index: 1.5640
Boiling Point: 183-185°
Beilstein Reference: 1071474
MDL No.: MFCD00009272
Formula:
C₆H₅CCCH₃ Boiling Pt: 183…185 °C Density: 0.933 Flash Pt: 62 °C(143 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009272 CAS Number: 673-32-5 EINECS: 211-607-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...