Benzoic anhydride 98%

Supplier: Thermo Scientific Chemicals
Danger

A14269-0B A14269-18 A14269-30
CAAAA14269-0BEA 461.36 CAD
CAAAA14269-0B CAAAA14269-18 CAAAA14269-30
Benzoic anhydride 98%
Benzoic anhydride

Purity: 98% CAS No.: 93-97-0 Molecular Formula: C14H10O3 Formula Weight: 226.23 Flash Point: 110°(230°F) Physical Form: Powder Density: 1.199 Sensitivity: Moisture Sensitive Odor: Odorless Melting Point: 40-44° Boiling Point: 360° Beilstein Reference: 516726 Merck Reference: 14,1092 MDL No.: MFCD00003073

Formula: C₁₄H₁₀O₃
Boiling Pt: 360 °C
Melting Pt: 40…44 °C
Density: 1.199
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003073
CAS Number: 93-97-0
EINECS: 202-291-1
Merck Index: 14,1092

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR