3,5-Dimethylbenzyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α-Bromomesitylene, 1-(Bromomethyl)-3,5-dimethylbenzene
A14267-06
A14267-14
CAAAA14267-06EA
84.21
CAD
CAAAA14267-06
CAAAA14267-14
CAAAA14267-22
3,5-Dimethylbenzyl bromide 98%
3,5-Dimethylbenzyl bromide
Purity: 98%
CAS No.: 27129-86-8
Molecular Formula: C9H11Br
Formula Weight: 199.10
Melting Point: 36-40°
Boiling Point: 109-110°/14mm
UN No.: 3261
MDL No.: MFCD00013539
Formula:
C₉H₁₁Br MW: 199.09 g/mol Boiling Pt: 232 Melting Pt: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013539 CAS Number: 27129-86-8 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...