Phenylacetaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14263-14
A14263-22
A14263-36
CAAAAA14263-14EA
37.92
CAD
CAAAAA14263-14
CAAAA14263-22
CAAAA14263-36
Phenylacetaldehyde 95%
Phenylacetaldehyde
Purity: 95%
CAS No.: 122-78-1
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5350
Boiling Point: 194-196°
Beilstein Reference: 385791
Merck Reference: 14,7265
MDL No.: MFCD00006993
Formula:
C₈H₈O Boiling Pt: 194…196 °C Melting Pt: 33…34 °C Density: 1.075 Flash Pt: 68 °C(154 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006993 CAS Number: 122-78-1 EINECS: 204-574-5 Merck Index: 14,7265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...