cis-2-Penten-1-ol 97% remainder mainly trans-isomer

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Z)-2-Penten-1-ol

A14262-06 A14262-14 A14262-22
CAAAA14262-06EA 65.1 CAD
CAAAA14262-06 CAAAA14262-14 CAAAA14262-22
cis-2-Penten-1-ol 97% remainder mainly trans-isomer
cis-2-Penten-1-ol

Purity: 97% CAS No.: 1576-95-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 48°(118°F) Refractive Index: 1.4360 Boiling Point: 138° Beilstein Reference: 1719473 UN No.: 1987 MDL No.: MFCD00063208

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 138 °C
Density: 0.853
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063208
CAS Number: 1576-95-0
EINECS: 216-415-7
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR