N-(2-Hydroxyethyl)phthalimide 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(2-Hydroxyethyl)isoindoline-1,3-dione
A14259-22
A14259-36
CAAAA14259-22EA
94.33
CAD
CAAAA14259-22
CAAAA14259-36
N-(2-Hydroxyethyl)phthalimide 98+%
N-(2-Hydroxyethyl)phthalimide
Purity: 98+%
CAS No.: 3891-07-4
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Melting Point: 126-129°
Beilstein Reference: 147214
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol Melting Pt: 126…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005903 CAS Number: 3891-07-4 EINECS: 223-434-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...