Dimethyl phthalate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phthalic acid dimethyl ester
A14253-30
A14253-0E
A14253-36
CAAAAA14253-30EA
36.59
CAD
CAAAAA14253-30
CAAAA14253-0E
CAAAA14253-36
Dimethyl phthalate 99%
Dimethyl phthalate
Purity: 99%
CAS No.: 131-11-3
Molecular Formula: C10H10O4
Formula Weight: 194.19
Flash Point: 146°(294°F)
Physical Form: Liquid
Density: 1.190
Refractive Index: 1.5150
Melting Point: ca 5°
Boiling Point: 283-284°
Beilstein Reference: 1911460
Merck Reference: 14,3255
MDL No.: MFCD00008425
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Boiling Pt: 283.7 Melting Pt: 6 Density: 1.190 Flash Pt: 146 °C (294 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008425 CAS Number: 131-11-3 EINECS: 205-011-6 Merck Index: 14,3255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...