3-Acetylpyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone

A14246-14 A14246-22 A14246-36
CAAAA14246-14EA 58.93 CAD
CAAAA14246-14 CAAAA14246-22 CAAAA14246-36
3-Acetylpyridine 98%
3-Acetylpyridine

Purity: 98% CAS No.: 350-03-8 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 104°(219°F) Refractive Index: 1.5350 Odor: Characteristic Melting Point: 12-13° Boiling Point: 218-220° Beilstein Reference: 107751 Merck Reference: 14,6116 UN No.: 2810 MDL No.: MFCD00006396

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 218…220 °C
Melting Pt: 12…13 °C
Density: 1.106
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 14,6116

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR