5-Chloro-o-anisidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-2-methoxyaniline, 5-Chloro-2-anisidine, 2-Amino-4-chloroanisole

A14244-22 A14244-36
CAAAAA14244-0EEA 2101.42 CAD
CAAAAA14244-0E CAAAA14244-22 CAAAA14244-36
5-Chloro-o-anisidine 98%
5-Chloro-o-anisidine

Purity: 98% CAS No.: 95-03-4 Molecular Formula: C7H8ClNO Formula Weight: 157.60 Odor: Characteristic Melting Point: 79-84° Boiling Point: 135°/8mm Beilstein Reference: 879161 UN No.: 2233 MDL No.: MFCD00007777

Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 135 °C/8 mm
Melting Pt: 79…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00007777
CAS Number: 95-03-4
EINECS: 202-385-2
UN: 2233
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR