4-Ethyl-2-methoxyphenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14239-06
A14239-14
CAAAA14239-06EA
98.75
CAD
CAAAA14239-06
CAAAA14239-14
CAAAA14239-22
4-Ethyl-2-methoxyphenol 98%
4-Ethyl-2-methoxyphenol
Purity: 98%
CAS No.: 2785-89-9
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 107°(225°F)
Refractive Index: 1.5280
Melting Point: 15°
Boiling Point: 230°
Beilstein Reference: 2045329
MDL No.: MFCD00038714
Formula:
C₉H₁₂O₂ Boiling Pt: 230 °C Melting Pt: 15 °C Density: 1,064 Flash Pt: 107 °C(225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038714 CAS Number: 2785-89-9 EINECS: 220-500-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...