Bis(2-chloroethyl)ether 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14234-0B
A14234-18
A14234-30
CAAAA14234-0BEA
456.62
CAD
CAAAA14234-0B
CAAAA14234-18
CAAAA14234-30
Bis(2-chloroethyl)ether 99%
Bis(2-chloroethyl) ether
Purity: 99%
CAS No.: 111-44-4
Molecular Formula: C4H8Cl2O
Formula Weight: 143.01
Flash Point: 55°(131°F)
Refractive Index: 1.4570
Odor: Pungent
Melting Point: -50°
Boiling Point: 178°
Beilstein Reference: 605317
Merck Reference: 14,3066
UN No.: 1916
MDL No.: MFCD00000975
Formula:
C₄H₈Cl₂O Boiling Pt: 178 °C Melting Pt: –50 °C Density: 1.219 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000975 CAS Number: 111-44-4 EINECS: 203-870-1 UN: 1916 ADR: 6.1,II Merck Index: 14,3066 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...