Ethyl chlorodifluoroacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

A14232-14 A14232-22
CAAAA14232-14EA 123.87 CAD
CAAAA14232-14 CAAAA14232-22
Ethyl chlorodifluoroacetate 98%
Ethyl chlorodifluoroacetate

Purity: 98% CAS No.: 383-62-0 Molecular Formula: ClF2CCO2CH2CH3 Formula Weight: 158.53 Flash Point: 18°(64°F) Refractive Index: 1.3580 Boiling Point: 96-97° Beilstein Reference: 1761413 UN No.: 2924 MDL No.: MFCD00013662

Formula: C₄H₅ClF₂O₂
Boiling Pt: 96…97 °C
Density: 1.252
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013662
CAS Number: 383-62-0
EINECS: 206-850-0
UN: 2924
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR