α,α,α-Trifluoro-o-cresol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxybenzotrifluoride, 2-(Trifluoromethyl)phenol

A14231-03 A14231-06 A14231-14
CAAAAA14231-03EA 73.15 CAD
CAAAAA14231-03 CAAAA14231-06 CAAAA14231-14
α,α,α-Trifluoro-o-cresol 98%
α,α,α-Trifluoro-o-cresol

Purity: 98% CAS No.: 444-30-4 Molecular Formula: C7H5F3O Formula Weight: 162.11 Flash Point: 65°(149°F) Odor: Phenol-like Boiling Point: 147-148° Beilstein Reference: 1867917 UN No.: 3261 MDL No.: MFCD00002222

Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 147…148 °C
Melting Pt: 45…46 °C
Density: Min. 1 g/cm³ (20 °C)
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR