(±)-Mandelic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A14223-0E A14223-22 A14223-36
CAAAA14223-0EEA 545.1 CAD
CAAAA14223-0E CAAAA14223-22 CAAAA14223-36
(±)-Mandelic acid 99%
(±)-Mandelic acid

Purity: 99% CAS No.: 90-64-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Physical Form: Cryst powder Sensitivity: Light Sensitive Appearance: White Melting Point: 118-121° Beilstein Reference: 510011 Merck Reference: 14,5717 MDL No.: MFCD00064250

Formula: C₈H₈O₃
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 118…121 °C
Density: 1.361 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064250
CAS Number: 90-64-2
EINECS: 202-007-6
Merck Index: 14,5717

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR