5-Bromo-2-fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14219-30
A14219-09
A14219-18
CAAAAA14219-30EA
1297.18
CAD
CAAAAA14219-30
CAAAA14219-09
CAAAA14219-18
5-Bromo-2-fluorobenzaldehyde 98%
5-Bromo-2-fluorobenzaldehyde
Purity: 98%
CAS No.: 93777-26-5
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Flash Point: 109°(228°F)
Refractive Index: 1.5700
Boiling Point: 230°
Beilstein Reference: 7632798
MDL No.: MFCD00070755
Formula:
C₇H₄BrFO Boiling Pt: 230 °C Melting Pt: 49 °C Density: 1,710 Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070755 CAS Number: 93777-26-5 EINECS: 298-056-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...