Diacetyl 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Butanedione

A14217-09 A14217-22 A14217-36
CAAAAA14217-09EA 59.41 CAD
CAAAAA14217-09 CAAAA14217-22 CAAAA14217-36
Diacetyl 99%
Diacetyl

Purity: 99% CAS No.: 431-03-8 Molecular Formula: C4H6O2 Formula Weight: 86.09 Flash Point: 10°(50°F) Refractive Index: 1.3940 Odor: Sweet Melting Point: -3° Boiling Point: 87-88° Beilstein Reference: 605398 Merck Reference: 14,2966 UN No.: 2346 MDL No.: MFCD00008756

Formula: C₄H₆O₂
Boiling Pt: 87…88 °C
Melting Pt: –3 °C
Density: 0.983
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008756
CAS Number: 431-03-8
EINECS: 207-069-8
UN: 2346
ADR: 3,II
Merck Index: 14,2966

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR