Cyclopropanecarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14215-0B
A14215-18
A14215-30
CAAAA14215-0BEA
674.66
CAD
CAAAA14215-0B
CAAAA14215-18
CAAAA14215-30
Cyclopropanecarboxylic acid 98%
Cyclopropanecarboxylic acid
Purity: 98%
CAS No.: 1759-53-1
Molecular Formula: C4H6O2
Formula Weight: 86.09
Flash Point: 71°(159°F)
Refractive Index: 1.4390
Melting Point: 5°
Boiling Point: 182-184°
Beilstein Reference: 969839
UN No.: 3265
MDL No.: MFCD00001287
Formula:
C₄H₆O₂ Boiling Pt: 182…184 °C Melting Pt: 5 °C Density: 1.086 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001287 CAS Number: 1759-53-1 EINECS: 217-162-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...