3,4-Diaminobenzoic acid 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14213-14
A14213-22
A14213-36
CAAAA14213-14EA
98.75
CAD
CAAAA14213-14
CAAAA14213-22
CAAAA14213-36
3,4-Diaminobenzoic acid 94%
3,4-Diaminobenzoic acid
Purity: 94%
CAS No.: 619-05-6
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: ca 210° dec.
Beilstein Reference: 775892
Formula:
C₇H₈N₂O₂ Boiling Pt: 354 °C (1013 hPa) Melting Pt: ca 210 °C (dec.) Density: 1.544 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007726 CAS Number: 619-05-6 EINECS: 210-577-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...