1,6-Diaminohexane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,6-Hexanediamine, Hexamethylenediamine
A14212-22
A14212-36
CAAAA14212-22EA
49.52
CAD
CAAAA14212-22
CAAAA14212-36
1,6-Diaminohexane 98+%
1,6-Diaminohexane
Purity: 98+%
CAS No.: 124-09-4
Molecular Formula: C6H16N2
Formula Weight: 116.21
Flash Point: 85°(185°F)
Odor: Amine-like
pH: 12.4
Melting Point: 39-43°
Boiling Point: 199-204°
Beilstein Reference: 1098307
Merck Reference: 14,4695
UN No.: 2280
MDL No.: MFCD00008243
Formula:
C₆H₁₆N₂ Boiling Pt: 199…204 °C Melting Pt: 39…43 °C Density: 0.840 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008243 CAS Number: 124-09-4 EINECS: 204-679-6 UN: 2280 ADR: 8,III Merck Index: 14,4695 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...