3-Iodoanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-iodoanisole

A14208-06 A14208-14 A14208-22
CAAAA14208-06EA 46.61 CAD
CAAAA14208-06 CAAAA14208-14 CAAAA14208-22
3-Iodoanisole 98%
3-Iodoanisole

Purity: 98% CAS No.: 766-85-8 Molecular Formula: C7H7IO Formula Weight: 234.04 Flash Point: 110°(230°F) Refractive Index: 1.6130 Boiling Point: 243-244° Beilstein Reference: 1858896 MDL No.: MFCD00001048

Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 243…244 °C
Density: 1.780
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001048
CAS Number: 766-85-8
EINECS: 212-173-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR