D-(+)-Biotin 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

A14207-09 A14207-03 A14207-06 A14207-14
CAAAAA14207-09EA 957.48 CAD
CAAAAA14207-09 CAAAA14207-03 CAAAA14207-06 CAAAA14207-14
D-(+)-Biotin 98+%
D(+)-Biotin

Essential vitamin that is important for amino acid and energy metabolism


Purity: 98+% CAS No.: 58-85-5 Molecular Formula: C10H16N2O3S Formula Weight: 244.32 Storage Temperature: 2°C to 8°C Melting Point: 224-226° Beilstein Reference: 86838 Merck Reference: 14,1231 Optical Rotation: +91° (c=1 in 0.1N NaOH) MDL No.: MFCD00005541


Essential vitamin that is important for amino acid and energy metabolism

Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Melting Pt: 224…226 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 14,1231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR