2-Chloro-5-nitrobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Chloro-5-nitrophenyl)methanol

A14205-06 A14205-14 A14205-22
CAAAA14205-06EA 50.24 CAD
CAAAA14205-06 CAAAA14205-14 CAAAA14205-22
2-Chloro-5-nitrobenzyl alcohol 98%
2-Chloro-5-nitrobenzyl alcohol

Purity: 98% CAS No.: 80866-80-4 Molecular Formula: C7H6ClNO3 Formula Weight: 187.58 Melting Point: 75-79° Beilstein Reference: 3248759

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00007297
CAS Number: 80866-80-4
EINECS: 279-584-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR