L(+)-Glutamine 99%
Supplier: Thermo Scientific Chemicals
A14201-0B
A14201-18
A14201-30
CAAAA14201-0BEA
695.2
CAD
CAAAA14201-0B
CAAAA14201-18
CAAAA14201-30
L(+)-Glutamine 99%
L(+)-Glutamine
Purity: 99%
CAS No.: 56-85-9
Molecular Formula: C5H10N2O3
Formula Weight: 146.15
Physical Form: Powder
Appearance: White
Melting Point: 185° dec.
Beilstein Reference: 1723797
Merck Reference: 14,4471
Optical Rotation: +33° (c=5 in 5N HCl)
MDL No.: MFCD00008044
Formula:
C₅H₁₀N₂O₃ Boiling Pt: 355 °C (1013 hPa) Melting Pt: 185 °C (dec.) Density: 1.5326 g/cm³ (130 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008044 CAS Number: 56-85-9 EINECS: 200-292-1 Merck Index: 14,4471 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...