2,4,5-Trifluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14193-14 A14193-06
CAAAAA14193-14EA 524.56 CAD
CAAAAA14193-14 CAAAA14193-03 CAAAA14193-06
2,4,5-Trifluorobenzoic acid 98%
2,4,5-Trifluorobenzoic acid

Purity: 98% CAS No.: 446-17-3 Molecular Formula: C7H3F3O2 Formula Weight: 176.10 Melting Point: 95-100° Beilstein Reference: 3257609

Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00013306
CAS Number: 446-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR