D-(-)-Glutamic acid 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid

A14191-22 A14191-06 A14191-14
CAAAAA14191-22EA 726.8 CAD
CAAAAA14191-22 CAAAA14191-06 CAAAA14191-14
D-(-)-Glutamic acid 99+%
D(-)-Glutamic acid

Purity: 99+% CAS No.: 6893-26-1 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Melting Point: 199° dec. Beilstein Reference: 1723800 Merck Reference: 14,4469 Optical Rotation: -30° (c=10 in 2N HCl) MDL No.: MFCD00063112

Formula: C₅H₉NO₄
MW: 147.13 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 199 °C (dec.)
Density: 1.533 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063112
CAS Number: 6893-26-1
EINECS: 230-000-8
Merck Index: 14,4469

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR