1,4-Dioxane 99+% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Stabilization: ∼ 5-10 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A14188-0E A14188-36
CAAAA14188-0EEA 222.78 CAD
CAAAA14188-0E CAAAA14188-36
1,4-Dioxane 99+% stabilised
1,4-Dioxane

Purity: 99+% CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Refractive Index: 1.4220 Odor: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1
Melting Pt: 11.8
Density: 1.034
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 14,3300
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR