2-Fluorobenzenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14181-06
A14181-14
CAAAA14181-06EA
140.94
CAD
CAAAA14181-06
CAAAA14181-14
2-Fluorobenzenesulfonyl chloride 98%
2-Fluorobenzenesulphonyl chloride
Purity: 98%
CAS No.: 2905-21-7
Molecular Formula: C6H4ClFO2S
Formula Weight: 194.61
Flash Point:
Formula:
C₆H₄ClFO₂S Boiling Pt: 246…247 °C Melting Pt: 27…32 °C Density: 1.470 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042285 CAS Number: 2905-21-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...