4-Bromo-3,5-dimethylisoxazole 97%

Supplier: Thermo Scientific Chemicals
Warning

A14179-03 A14179-06 A14179-14
CAAAA14179-03EA 65.73 CAD
CAAAA14179-03 CAAAA14179-06 CAAAA14179-14
4-Bromo-3,5-dimethylisoxazole 97%
4-Bromo-3,5-dimethylisoxazole

Purity: 97% CAS No.: 10558-25-5 Molecular Formula: C5H6BrNO Formula Weight: 176.02 Storage Temperature: 2°C to 8°C Flash Point: 73°(163°F) Refractive Index: 1.4880 Boiling Point: 176° Beilstein Reference: 108522 MDL No.: MFCD00068187

Formula: C₅H₆BrNO
MW: 176.01 g/mol
Boiling Pt: 176 °C
Density: 1,49
Flash Pt: 73 °C(163 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00068187
CAS Number: 10558-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR