2-Amino-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine
A14177-14
A14177-22
A14177-36
CAAAAA14177-14EA
41.4
CAD
CAAAAA14177-14
CAAAA14177-22
CAAAA14177-36
2-Amino-4-methylpyridine 98%
2-Amino-4-methylpyridine
Purity: 98%
CAS No.: 695-34-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 118°(244°F)
Odor: Unpleasant
Boiling Point: 231°
Beilstein Reference: 107066
Merck Reference: 14,466
UN No.: 2811
MDL No.: MFCD00006321
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 231 °C Melting Pt: 97…101 °C Flash Pt: 118 °C (244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006321 CAS Number: 695-34-1 EINECS: 211-780-9 UN: 2811 ADR: 6.1,III Merck Index: 14,466 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...