α,α,α-Trifluoro-p-toluoyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14176-06
A14176-14
CAAAA14176-06EA
98.91
CAD
CAAAA14176-06
CAAAA14176-14
CAAAA14176-22
α,α,α-Trifluoro-p-toluoyl chloride 97%
Purity: 97%
CAS No.: 329-15-7
Molecular Formula: C8H4ClF3O
Formula Weight: 208.57
Flash Point: 78°(172°F)
Refractive Index: 1.4770
Odor: Pungent
Melting Point: -3°
Boiling Point: 188-190°
Beilstein Reference: 391282
UN No.: 3265
MDL No.: MFCD00000694
Formula:
C₈H₄ClF₃O Boiling Pt: 188…190 °C Melting Pt: –3 °C Density: 1.404 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000694 CAS Number: 329-15-7 EINECS: 206-342-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...