N,N-Dimethyl-p-phenylenediammonium sulfate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14175-36
A14175-14
A14175-22
CAAAAA14175-36EA
1527.86
CAD
CAAAAA14175-36
CAAAA14175-14
CAAAA14175-22
N,N-Dimethyl-p-phenylenediammonium sulfate 98%
N,N-Dimethyl-p-phenylenediammonium sulphate
Purity: 98%
CAS No.: 536-47-0
Molecular Formula: C8H12N2.H2SO4
Formula Weight: 234.28
Odor: Odorless
Melting Point: 200-205° dec.
Beilstein Reference: 4612278
UN No.: 2811
MDL No.: MFCD00012992
Formula:
C₈H₁₄N₂O₄S Melting Pt: 200…205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012992 CAS Number: 536-47-0 EINECS: 208-636-2 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...