3-Chloro-6-methoxypyridazine 97%

Supplier: Thermo Scientific Chemicals
Warning

A14173-22 A14173-06 A14173-14
CAAAAA14173-22EA 1243.46 CAD
CAAAAA14173-22 CAAAA14173-06 CAAAA14173-14
3-Chloro-6-methoxypyridazine 97%
3-Chloro-6-methoxypyridazine

Purity: 97% CAS No.: 1722-10-7 Molecular Formula: C5H5ClN2O Formula Weight: 144.56 Melting Point: 90-92° Beilstein Reference: 118854

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
Boiling Pt: 174 - 175 °C (3 -rr)
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006467
CAS Number: 1722-10-7
EINECS: 217-019-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR