4-Hexylbenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14168-03
A14168-06
CAAAA14168-03EA
91.8
CAD
CAAAA14168-03
CAAAA14168-06
4-Hexylbenzoic acid 99%
4-Hexylbenzoic acid
Purity: 99%
CAS No.: 21643-38-9
Molecular Formula: C13H18O2
Formula Weight: 206.29
Melting Point: (C-N) 97-99°
Beilstein Reference: 2046454
Formula:
C₁₃H₁₈O₂ MW: 206.28 g/mol Boiling Pt: 330 °C (1013 hPa) Melting Pt: 97…99 °C (C-Nematic) Storage Temperature: Ambient |
MDL Number:
MFCD00013997 CAS Number: 21643-38-9 EINECS: 244-490-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...