3,4,5-Triethoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14157-14 A14157-22 A14157-36
CAAAA14157-14EA 46.14 CAD
CAAAA14157-14 CAAAA14157-22 CAAAA14157-36
3,4,5-Triethoxybenzoic acid 98+%
3,4,5-Triethoxybenzoic acid

Purity: 98+% CAS No.: 6970-19-0 Molecular Formula: C13H18O5 Formula Weight: 254.29 Melting Point: 110-113° Beilstein Reference: 2659063

Formula: C₁₃H₁₈O₅
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002505
CAS Number: 6970-19-0
EINECS: 230-192-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR