1,3-Dichloroacetone 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14149-14
A14149-22
A14149-36
CAAAA14149-14EA
72.52
CAD
CAAAA14149-14
CAAAA14149-22
CAAAA14149-36
1,3-Dichloroacetone 96%
1,3-Dichloroacetone
Purity: 96%
CAS No.: 534-07-6
Molecular Formula: C3H4Cl2O
Formula Weight: 126.97
Storage Temperature: 2°C to 8°C
Flash Point: 95°(203°F)
Refractive Index: 1.4400
Odor: Pungent
pH: 4.2
Melting Point: 40-44°
Boiling Point: 172-173°
Beilstein Reference: 605456
Merck Reference: 14,3052
UN No.: 2649
MDL No.: MFCD00000937
Formula:
ClH₂CC(O)CH₂Cl Boiling Pt: 172…173 °C Melting Pt: 40…44 °C Density: 1.383 Flash Pt: 95 °C (203 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000937 CAS Number: 534-07-6 EINECS: 208-585-6 UN: 2649 ADR: 6.1,II Merck Index: 14,3052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...