2-Methylpyrazine 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14146-14
A14146-22
A14146-36
CAAAA14146-14EA
48.66
CAD
CAAAA14146-14
CAAAA14146-22
CAAAA14146-36
2-Methylpyrazine 99+%
2-Methylpyrazine
Purity: 99+%
CAS No.: 109-08-0
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 50°(122°F)
Refractive Index: 1.5050
Odor: Unpleasant
Melting Point: -30°
Boiling Point: 135-136°
Beilstein Reference: 105778
UN No.: 1993
MDL No.: MFCD00006142
Formula:
C₅H₆N₂ MW: 94.12 g/mol Boiling Pt: 135…136 °C Melting Pt: –30 °C Density: 1.028 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006142 CAS Number: 109-08-0 EINECS: 203-645-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...