2-Decanone 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Decan-2-one
A14145-06
A14145-14
A14145-22
CAAAAA14145-06EA
46.93
CAD
CAAAAA14145-06
CAAAA14145-14
CAAAA14145-22
2-Decanone 97%
2-Decanone
Purity: 97%
CAS No.: 693-54-9
Molecular Formula: CH3(CH2)7COCH3
Formula Weight: 156.27
Flash Point: 86°(187°F)
Refractive Index: 1.4250
Melting Point: 3°
Boiling Point: 210-212°
Beilstein Reference: 1747463
MDL No.: MFCD00009571
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Boiling Pt: 211 Melting Pt: 3 Density: 0.824 Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009571 CAS Number: 693-54-9 EINECS: 211-752-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...