2-(4-Bromophenyl)ethanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14143-03
A14143-06
A14143-14
CAAAA14143-03EA
44.71
CAD
CAAAA14143-03
CAAAA14143-06
CAAAA14143-14
2-(4-Bromophenyl)ethanol 98%
2-(4-Bromophenyl)ethanol
Purity: 98%
CAS No.: 4654-39-1
Molecular Formula: C8H9BrO
Formula Weight: 201.06
Flash Point:
Formula:
C₈H₉BrO Boiling Pt: 106…108 °C/1 mm Melting Pt: 36…38 °C Density: 1.436 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002897 CAS Number: 4654-39-1 EINECS: 225-093-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...