Cyclopentanecarbonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14142-06
A14142-14
A14142-22
CAAAA14142-06EA
107.91
CAD
CAAAA14142-06
CAAAA14142-14
CAAAA14142-22
Cyclopentanecarbonyl chloride 98%
Cyclopentanecarbonyl chloride
Purity: 98%
CAS No.: 4524-93-0
Molecular Formula: C6H9ClO
Formula Weight: 132.59
Flash Point: 60°(140°F)
Refractive Index: 1.4640
Odor: Acrid
Boiling Point: 160-162°
Beilstein Reference: 1237147
UN No.: 2920
MDL No.: MFCD00001370
Formula:
C₆H₉ClO Boiling Pt: 160…162 °C Density: 1.091 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001370 CAS Number: 4524-93-0 EINECS: 224-856-1 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...