4-Fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14129-14 A14129-22 A14129-36
CAAAA14129-14EA 54.83 CAD
CAAAA14129-14 CAAAA14129-22 CAAAA14129-36
4-Fluorobenzoic acid 98%
4-Fluorobenzoic acid

Purity: 98% CAS No.: 456-22-4 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 183-187° Beilstein Reference: 1906922 Merck Reference: 14,4171 MDL No.: MFCD00002530

Formula: C₇H₅FO₂
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 183…187 °C
Density: 1.470
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0
Merck Index: 14,4171

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR