1-Bromonaphthalene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
α-Bromonaphthalene
A14122-0B
A14122-18
CAAAAA14122-09EA
45.69
CAD
CAAAAA14122-09
CAAAA14122-0B
CAAAA14122-18
1-Bromonaphthalene 97%
1-Bromonaphthalene
Purity: 97%
CAS No.: 90-11-9
Molecular Formula: C10H7Br
Formula Weight: 207.08
Flash Point:
Formula:
C₁₀H₇Br MW: 207.07 g/mol Boiling Pt: 279…281 °C Melting Pt: –2 °C…0 °C Density: 1.485 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003868 CAS Number: 90-11-9 EINECS: 201-965-2 Merck Index: 14,1425 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...