α,α,α',α'-Tetrabromo-o-xylene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Bis(dibromomethyl)benzene

A14121-18 A14121-30
CAAAA14121-18EA 118.82 CAD
CAAAA14121-18 CAAAA14121-30
α,α,α',α'-Tetrabromo-o-xylene 97%
α,α,α',α'-Tetrabromo-o-xylene

Purity: 97% CAS No.: 13209-15-9 Molecular Formula: C8H6Br4 Formula Weight: 421.75 Melting Point: 114-118° Beilstein Reference: 2211698 UN No.: 1759 MDL No.: MFCD00000131

Formula: C₈H₆Br₄
MW: 421.75 g/mol
Boiling Pt: 346
Melting Pt: 117
Storage Temperature: Ambient
MDL Number: MFCD00000131
CAS Number: 13209-15-9
EINECS: 236-176-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR