2-Bromophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14112-14
A14112-22
A14112-36
CAAAA14112-14EA
186.44
CAD
CAAAA14112-14
CAAAA14112-22
CAAAA14112-36
2-Bromophenol 98%
2-Bromophenol
Purity: 98%
CAS No.: 95-56-7
Molecular Formula: C6H5BrO
Formula Weight: 173.01
Flash Point: 89°(192°F)
Refractive Index: 1.5890
Odor: Phenol-like
Melting Point: 5°
Boiling Point: 194-195°
Beilstein Reference: 1905115
Merck Reference: 14,1428
UN No.: 3082
MDL No.: MFCD00002146
Formula:
C₆H₅BrO Boiling Pt: 194…195 °C Melting Pt: 5 °C Density: 1.636 Flash Pt: 92 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002146 CAS Number: 95-56-7 EINECS: 202-432-7 UN: 1993 ADR: 3,III Merck Index: 14,1428 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...