2-(Trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Trifluoromethyl)benzene-1-sulphonyl chloride, 2-(Trifluoromethyl)benzene-1-sulfonyl chloride

A14108-03 A14108-22 A14108-14
CAAAAA14108-03EA 48.98 CAD
CAAAAA14108-03 CAAAAA14108-22 CAAAA14108-14
2-(Trifluoromethyl)benzenesulfonyl chloride 98%
2-(Trifluoromethyl)benzenesulphonyl chloride

Purity: 98% CAS No.: 776-04-5 Molecular Formula: C7H4ClF3O2S Formula Weight: 244.62 Flash Point: 110°(230°F) Refractive Index: 1.5030 Melting Point: 22-24° Boiling Point: 133-135°/14mm Beilstein Reference: 2651854 UN No.: 3261 MDL No.: MFCD00051696

Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 133…135 °C (14 mmHg)
Melting Pt: 22…24 °C
Density: 1.585
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR