1,4-Dimethoxynaphthalene 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14095-03
A14095-06
A14095-14
CAAAA14095-03EA
118.97
CAD
CAAAA14095-03
CAAAA14095-06
CAAAA14095-14
1,4-Dimethoxynaphthalene 96%
1,4-Dimethoxynaphthalene
Purity: 96%
CAS No.: 10075-62-4
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: 84-85°
Beilstein Reference: 2048082
Formula:
C₁₂H₁₂O₂ Melting Pt: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052378 CAS Number: 10075-62-4 EINECS: 233-209-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...