Triphenylphosphine 99%, flakes

Supplier: Thermo Scientific Chemicals
Warning

A14089-0B A14089-0I A14089-30
CAAAA14089-0BEA 274.92 CAD
CAAAA14089-0B CAAAA14089-0I CAAAA14089-30
Triphenylphosphine 99%, flakes
Triphenylphosphine

In the synthesis of organic compounds, phosphonium salts and other phosphorus compounds, and as a polymerization initiator


Purity: 99% CAS No.: 603-35-0 Molecular Formula: (C6H5)3P Formula Weight: 262.29 Flash Point: 181°(358°F) Physical Form: Flakes Density: 1.200 Appearance: White to pale cream Odor: Mild Melting Point: 79-81° Boiling Point: 360° Solubility: Insoluble in water. Soluble in benzene, acetone, carbon tetrachloride, ether, and glacial acetic acid Beilstein Reference: 610776 Merck Reference: 14,9743 MDL No.: MFCD00003043


In the synthesis of organic compounds, phosphonium salts and other phosphorus compounds, and as a polymerization initiator

Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 360 °C
Melting Pt: 79…81 °C
Density: 1.200
Flash Pt: 181 °C(358 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 14,9743

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR