4-Aminothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline

A14082-06 A14082-14 A14082-22
CAAAA14082-06EA 91.48 CAD
CAAAA14082-06 CAAAA14082-14 CAAAA14082-22
4-Aminothiophenol 97%
4-Aminothiophenol

Purity: 97% CAS No.: 1193-02-8 Molecular Formula: C6H7NS Formula Weight: 125.19 Flash Point: 110°(230°F) Odor: Stench Boiling Point: 156-157° Beilstein Reference: 906904 UN No.: 3263 MDL No.: MFCD00007880

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 262
Melting Pt: 37…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007880
CAS Number: 1193-02-8
EINECS: 214-763-4
UN: 3263
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR